Läs mer
4-O-(E)-Feruloylquinic Acid (~90%), TRC
Produktkod. 30007065
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007065 5 mg
30004313 1 mg
30005346 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30007065 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF3089855MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 96646-16-1
Molekylformel C17H20O9
Molekylvikt (g/mol) 368.34
Synonym [1R-[1α,3α,4α(E),5β]]-1,3,5-Trihydroxy-4-[[3-(4-hydroxy-3-methoxyphenyl)-1-oxo-2-propenyl]oxy]-Cyclohexanecarboxylic Acid;
LEDER O[C@]1(C(O)=O)C[C@@H](O)[C@H](OC(/C=C/C2=CC=C(O)C(OC)=C2)=O)[C@H](O)C1
Specifikationer
Kemiskt namn eller material 4-O-(E)-Feruloylquinic Acid
Kvantitet 5 mg
InChI formel InChI=1S/C17H20O9/c1-25-13-6-9(2-4-10(13)18)3-5-14(21)26-15-11(19)7-17(24,16(22)23)8-12(15)20/h2-6,11-12,15,18-20,24H,7-8H2,1H3,(H,22,23)/b5-3+/t11-,12-,15?,17?/m1/s1