Läs mer
4-O-Methyl-D-glucuronic Acid, TRC
Produktkod. 30179059
Change view
Kvantitet:
1 mg
2.5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179059 1 mg
30179255 2.5 mg
30179048 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179059 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3228081MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2463-49-2
Molekylformel C7 H12 O7
Molekylvikt (g/mol) 208.17
Synonym 4-O-Methylglucuronic Acid
IUPAC-namn (2S,3S,4R,5R)-2,4,5-trihydroxy-3-methoxy-6-oxohexanoic acid
LEDER CO[C@@H]([C@H](O)[C@@H](O)C=O)[C@H](O)C(=O)O
Specifikationer
Kemiskt namn eller material 4-O-Methyl-D-glucuronic Acid linear
Kvantitet 1 mg
InChI formel InChI=1S/C7H12O7/c1-14-6(5(11)7(12)13)4(10)3(9)2-8/h2-6,9-11H,1H3,(H,12,13)/t3-,4+,5-,6-/m0/s1
Formel vikt 208.0583