Läs mer
4'-O-Methyl Quercetin, TRC
Product Code. 30006390
Change view
Click to view available options
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30006390 10 mg
30009283 5 mg
30007492 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30006390 Supplier Toronto Research Chemicals Supplier No. TRCM32658010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 603-61-2
Molekylformel C16 H12 O7
Molekylvikt (g/mol) 316.26
Synonym 3,5,7-Trihydroxy-2-(3-hydroxy-4-methoxyphenyl)-4H-1-benzopyran-4-one, 3,3',5,7-Tetrahydroxy-4'-methoxyflavone, Tamaraxetin, 4'-Methoxyquercetin, 4'-Methylquercetin, Quercetin 4'-methyl ether, Tamarixetin;
IUPAC-namn 3,5,7-trihydroxy-2-(3-hydroxy-4-methoxyphenyl)chromen-4-one
LEDER COc1ccc(cc1O)C2=C(O)C(=O)c3c(O)cc(O)cc3O2
Specifikationer
Kemiskt namn eller material Tamarixetin
Kvantitet 10 mg
InChI formel InChI=1S/C16H12O7/c1-22-11-3-2-7(4-9(11)18)16-15(21)14(20)13-10(19)5-8(17)6-12(13)23-16/h2-6,17-19,21H,1H3
Formel vikt 316.0583