Läs mer
4-Octyl Itaconate, MedChemExpress
Produktkod. 16491016
Encompass_Preferred
Change view
Kvantitet:
50 mg
100 mg
200 mg
500 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16491016 200 mg
16411616 500 mg
16425516 100 mg
16440967 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16491016 Leverantör Medchem Express Leverantörsnummer HY112675
MedChemExpress 4-Octyl Itaconate is a cell-permeable Itaconate derivative. Itaconate is an anti-inflammatory metabolite that activates Nrf2 via alkylation of KEAP1.
4-Octyl Itaconate is a cell-permeable Itaconate derivative. Itaconate is an anti-inflammatory metabolite that activates Nrf2 via alkylation of KEAP1.
Specifikationer
Kemiskt namn eller material 4-Octyl Itaconate
CAS 3133-16-2
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C13H22O4
Kvantitet 200 mg
Löslighetsinformation DMSO : 100 mg/mL (412.69 mM; Need ultrasonic)
LEDER O=C(OCCCCCCCC)CC(C(O)=O)=C
Molekylvikt (g/mol) 242.31
Formel vikt 242.31
Procent renhet 98.13%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) COVID-19-immunoregulation