Läs mer
4-Octylphenol Diethoxylate, TRC
Produktkod. 30138329
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138329 500 mg
17934775 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30138329 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO293765500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 51437-90-2
Molekylformel C18 H30 O3
Molekylvikt (g/mol) 294.43
Synonym 2-[2-(4-Octylphenoxy)ethoxy]ethanol, 2-[2-(p-Octylphenoxy)ethoxy]ethanol, 4-Octylphenol diethoxylate
IUPAC-namn 2-[2-(4-octylphenoxy)ethoxy]ethanol
LEDER CCCCCCCCc1ccc(OCCOCCO)cc1
Specifikationer
Kemiskt namn eller material 4-Octylphenol-di-ethoxylate
Kvantitet 500 mg
InChI formel InChI=1S/C18H30O3/c1-2-3-4-5-6-7-8-17-9-11-18(12-10-17)21-16-15-20-14-13-19/h9-12,19H,2-8,13-16H2,1H3
Formel vikt 294.2195