Läs mer
4-Oxo-4-(3-pyridyl)butanal, TRC
Produktkod. 30011986
Change view
Kvantitet:
1 mg
2.5 mg
10 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30011986 10 mg -
30004378 1 mg -
30004919 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011986 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO85925010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 76014-80-7
Molekylformel C9H9NO2
Molekylvikt (g/mol) 163.17
Synonym gamma-Oxo-3-pyridinebutanal, 4-Oxo-1-(3-pyridyl)-1-butanone;
IUPAC-namn 4-oxo-4-pyridin-3-ylbutanal
LEDER O=CCCC(=O)c1cccnc1
Specifikationer
Kemiskt namn eller material 4-Oxo-4-(3-pyridyl)butanal
Kvantitet 10 mg
InChI formel InChI=1S/C9H9NO2/c11-6-2-4-9(12)8-3-1-5-10-7-8/h1,3,5-7H,2,4H2
Formel vikt 163.0633