Läs mer
4-Oxopentanoic-13C3 Acid, TRC
Produktkod. 30410968
Change view
Kvantitet:
2.5 mg
Produktkod. Kvantitet
30410968 2.5 mg
1 options
Produktkod. 30410968 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO8599222.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Oxopentanoic-13C3 Acid
CAS 1391051-93-6
Molekylformel C213C3H8O3
Kvantitet 2.5 mg
Synonym 3-Acetylpropionic-13C3 Acid, 3-Oxobutanecarboxylic-13C3 Acid, 4-Ketovaleric-13C3 Acid, 4-Oxopentanoic-13C3 Acid, 4-Oxovaleric-13C3 Acid, Laevulinic-13C3 Acid, Levulic-13C3 Acid, NSC 3716-13C3, β-Acetylpropionic-13C3 Acid, γ-Ketovaleric-13C3 Acid, Levulinic-13C3 Acid
InChI formel InChI=1S/C5H8O3/c1-4(6)2-3-5(7)8/h2-3H2,1H3,(H,7,8)/i1+1,2+1,4+1
LEDER [13CH3][13C]([13CH2]CC(O)=O)=O
Molekylvikt (g/mol) 119.09
Formel vikt 119.0574