Läs mer
4-(p-Nitrophenyl)-3-butene-2-one, TRC
Produktkod. 30480714
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30480714 100 mg
1 options
Produktkod. 30480714 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN502220100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-(4-Nitrophenyl)-3-buten-2-one
CAS 3490-37-7
Molekylformel C10 H9 N O3
Kvantitet 100 mg
Synonym 3-Buten-2-one','4-(4-nitrophenyl)-, 3-Buten-2-one','4-(p-nitrophenyl)- (6CI,7CI,8CI), 4-(4-Nitrophenyl)-3-buten-2-one, 4-(p-Nitrophenyl)-3-buten-2-one, 4-Nitrobenzalacetone, Methyl 4-nitrostyryl ketone, Methyl p-nitrostyryl ketone, NSC 68341, p-Nitrobenzalacetone, p-Nitrobenzylideneacetone
InChI formel InChI=1S/C10H9NO3/c1-8(12)2-3-9-4-6-10(7-5-9)11(13)14/h2-7H,1H3
LEDER CC(=O)C=Cc1ccc(cc1)[N+](=O)[O-]
IUPAC-namn 4-(4-nitrophenyl)but-3-en-2-one
Molekylvikt (g/mol) 191.18
Formel vikt 191.0582