Läs mer
4-Penten-1-amine Hydrochloride, TRC
Produktkod. 30179587
Change view
Kvantitet:
1 g
2.5 g
Förpackningsstorlek:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30179587 1 g -
30179022 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30179587 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP2274031G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 27546-60-7
Molekylformel C5H11N • HCl
Molekylvikt (g/mol) 85.153646
Synonym 4-Pentenylamine Hydrochloride, 1-Amino-4-pentene Hydrochloride, 4-Penten-1-ylamine Hydrochloride, 4-Pentenamine Hydrochloride, 5-Aminopent-1-ene Hydrochloride
IUPAC-namn pent-4-en-1-amine;hydrochloride
LEDER C=CCCCN.Cl
Specifikationer
Kemiskt namn eller material 4-Penten-1-amine Hydrochloride
Kvantitet 1 g
InChI formel InChI=1S/C5H11N.ClH/c1-2-3-4-5-6;/h2H,1,3-6H2;1H
Formel vikt 121.0658