missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Pentenal, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
4-Pentenal is used in the preparation of 6-octen-3-one by addition with ethylene using 2,4-pentanedionatobis(ethylene)rhodium(I) as a catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Pentenal is used in the preparation of 6-octen-3-one by addition with ethylene using 2,4-pentanedionatobis(ethylene)rhodium(I) as a catalyst.
Solubility
Immiscible with water.
Notes
Air sensitive. Incompatible with strong oxidizing agents.
4-Pentenal is used in the preparation of 6-octen-3-one by addition with ethylene using 2,4-pentanedionatobis(ethylene)rhodium(I) as a catalyst.
Solubility
Immiscible with water.
Notes
Air sensitive. Incompatible with strong oxidizing agents.
Specifications
Density | 0.852 |
Boiling Point | 101°C to 105°C |
Flash Point | 7°C (44°F) |
Linear Formula | H2C=CH(CH2)2CHO |
Refractive Index | 1.417 |
Quantity | 1 g |
UN Number | UN1989 |
Beilstein | 1734369 |
Sensitivity | Air Sensitive |
Solubility Information | Immiscible with water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'