Läs mer
4-Pentynyl p-Tosylate, 90%, TRC
Produktkod. 30411981
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30411981 1 g
30421413 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30411981 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP2850501G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 77758-50-0
Molekylformel C12 H14 O3 S
Molekylvikt (g/mol) 238.3
Synonym 4-Pentynyl P-Tosylate
IUPAC-namn pent-4-ynyl 4-methylbenzenesulfonate
LEDER Cc1ccc(cc1)S(=O)(=O)OCCCC#C
Specifikationer
Kemiskt namn eller material 4-Pentynyl p-Tosylate
Kvantitet 1 g
InChI formel InChI=1S/C12H14O3S/c1-3-4-5-10-15-16(13,14)12-8-6-11(2)7-9-12/h1,6-9H,4-5,10H2,2H3
Formel vikt 238.0664