missing translation for 'onlineSavingsMsg'
Learn More

(4-Phenoxyphenyl)methylamine hydrochloride, 97%, Thermo Scientific™

Catalog Number p-4511328
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
169944-04-1
C13H14ClNO
235.711
VHCSCKHIGGFTHN-UHFFFAOYSA-N
4-phenoxyphenyl methanamine hydrochloride, 4-phenoxyphenyl methylamine hydrochloride, 4-phenoxybenzylamine hydrochloride, benzenemethanamine, 4-phenoxy-, hydrochloride, 1-4-phenoxyphenyl methanamine hydrochloride, 4-phenoxyphenyl methylamine, chloride, 1-4-phenoxyphenyl methanamine-hydrogen chloride 1/1
22293026
(4-phenoxyphenyl)methanamine;hydrochloride
C1=CC=C(C=C1)OC2=CC=C(C=C2)CN.Cl
This item is not returnable. View return policy
169944-04-1
C13H14ClNO
235.711
VHCSCKHIGGFTHN-UHFFFAOYSA-N
4-phenoxyphenyl methanamine hydrochloride, 4-phenoxyphenyl methylamine hydrochloride, 4-phenoxybenzylamine hydrochloride, benzenemethanamine, 4-phenoxy-, hydrochloride, 1-4-phenoxyphenyl methanamine hydrochloride, 4-phenoxyphenyl methylamine, chloride, 1-4-phenoxyphenyl methanamine-hydrogen chloride 1/1
22293026
(4-phenoxyphenyl)methanamine;hydrochloride
C1=CC=C(C=C1)OC2=CC=C(C=C2)CN.Cl
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 235.71
Percent Purity 97%
Chemical Name or Material (4-Phenoxyphenyl)methylamine hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'