Läs mer
4-Phenyl-1,2,4-triazoline-3,5-dione, TRC
Produktkod. 30416235
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30416235 1 g
1 options
Produktkod. 30416235 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP3373801G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Phenyl-1,2,4-triazoline-3,5-dione
CAS 4233-33-4
Molekylformel C8 H5 N3 O2
Kvantitet 1 g
Synonym Δ1-1,2,4-Triazoline-3,5-dione','4-phenyl- (7CI), 4-Phenyl-3H-1,2,4-triazole-3,5(4H)-dione, 4-Phenyl-1,2,4-triazole-3,5-dione, 4-Phenyl-1,2,4-triazoline-3,5-dione, 4-Phenyl-1,2,4-triazolinedione, 4-Phenyl-3,5-dioxotriazole, 4-Phenyl-3H-1,2,4-triazoline-3,5-dione, 4-Phenyl-4,5-dihydro-3H-1,2,4-triazole-3,5-dione, 4-Phenyl-4H-1,2,4-triazoline-3,5-dione, 4-Phenyl-4H-[1,2,4]triazole-3,5-dione, 4-Phenyl-Δ1-1,2,4-triazoline-3,5-dione, 4-Phenyldehydrourazole, 4-Phenyltriazoline-3,5-dione, Azodicarboxylic acid N-phenylimide, N-Phenyl-1,2,4-triazoline-3,5-dione, N-Phenyl-1,3,4-triazoline-2,5-dione, N-Phenyl-1,3,5-triazoline-2,4-dione, N-Phenylazodicarboximide, N-Phenylazodicarboxylic imide, N-Phenyltriazoline-3,5-dione, N-Phenyltriazolinedione, NSC 150362, PTAD
InChI formel InChI=1S/C8H5N3O2/c12-7-9-10-8(13)11(7)6-4-2-1-3-5-6/h1-5H
LEDER O=C1N=NC(=O)N1c2ccccc2
IUPAC-namn 4-phenyl-1,2,4-triazole-3,5-dione
Molekylvikt (g/mol) 175.14
Formel vikt 175.0382