Läs mer
4-phenyl-1,3-dioxolan-2-one, TRC
Produktkod. 30425260
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30425260 50 mg
30419928 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30425260 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP32206550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-phenyl-1,3-dioxolan-2-one
CAS 4427-92-3
Molekylformel C9H8O3
Kvantitet 50 mg
InChI formel InChI=1S/C9H8O3/c10-9-11-6-8(12-9)7-4-2-1-3-5-7/h1-5,8H,6H2
LEDER O=C1OCC(O1)c2ccccc2
IUPAC-namn 4-phenyl-1,3-dioxolan-2-one
Molekylvikt (g/mol) 164.15
Formel vikt 164.0473