Läs mer
4-Phenyl-1-(4-piperidinyl)-4-imidazolin-2-one, 96%, Thermo Scientific™
Produktkod. 11494830
Change view
Kvantitet:
250 mg
1 g
Förpackningsstorlek:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11494830 250 mg 250mg
11404840 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 11494830 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H50828.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 205058-28-2
Molekylformel C14H17N3O
Molekylvikt (g/mol) 243.31
MDL-nummer MFCD08061015
InChI-nyckel OPCVKWIYGHICKG-UHFFFAOYSA-N
Synonym 4-phenyl-1-4-piperidinyl-4-imidazolin-2-one, 4-phenyl-1-piperidin-4-yl-1,3-dihydro-2h-imidazol-2-one, 4-phenyl-1-piperidin-4-yl-1h-imidazol-2 3h-one, 4-phenyl-1-piperidin-4-yl-3h-imidazol-2-one, 4-phenyl-1-piperidin-4-yl-2,3-dihydro-1h-imidazol-2-one, 4-phenyl-1-4-piperidyl-4-imidazolin-2-one, acmc-1chii, 4-phenyl-piperidin-4-yl-1h-imidazol-2 3h-one, 1,3-dihydro-1-4-piperidinyl-4-phenyl-2 2h-imidazolone, 2h-imidazol-2-one, 1,3-dihydro-4-phenyl-1-4-piperidinyl
PubChem CID 11345546
IUPAC-namn 5-phenyl-3-piperidin-4-yl-1H-imidazol-2-one
LEDER C1CNCCC1N2C=C(NC2=O)C3=CC=CC=C3
Specifikationer
Kemiskt namn eller material 4-Phenyl-1-(4-piperidinyl)-4-imidazolin-2-one
Smältpunkt ∽235°C (decomposition)
Kvantitet 250 mg
Formel vikt 243.31
Procent renhet 96%