missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-1-butyne, 97%, Thermo Scientific Chemicals

Catalog Number 15161329
Click to view available options
Quantity:
5 g
25 g
16520-62-0
C10H10
130.19
QDEOKXOYHYUKMS-UHFFFAOYSA-N
4-phenyl-1-butyne, 3-butynylbenzene, but-3-yn-1-ylbenzene, benzene, 3-butynyl, 2-phenylethylacetylene, ccris 7100, phenethylacetylene, 4-phenyl-1-butyn, 3-butynylbenzene #, 4-phenylbut-1-yne
123360
but-3-ynylbenzene
C#CCCC1=CC=CC=C1
16520-62-0
C10H10
130.19
QDEOKXOYHYUKMS-UHFFFAOYSA-N
4-phenyl-1-butyne, 3-butynylbenzene, but-3-yn-1-ylbenzene, benzene, 3-butynyl, 2-phenylethylacetylene, ccris 7100, phenethylacetylene, 4-phenyl-1-butyn, 3-butynylbenzene #, 4-phenylbut-1-yne
123360
but-3-ynylbenzene
C#CCCC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.9260g/mL
Boiling Point 190°C
Flash Point 66°C
Assay Percent Range 96% min. (GC)
Quantity 25 g
Specific Gravity 0.926
Solubility Information Solubility in water: insoluble
Formula Weight 130.19
Percent Purity 97%
Chemical Name or Material 4-Phenyl-1-butyne
compliance-icons
Product Identifier
  • 4-Phenyl-1-butyne
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'