Läs mer
4-phenyl-2,3-dihydroquinazolin-2-one, TRC
Produktkod. 30417596
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30417596 100 mg
30410245 50 mg
30415192 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30417596 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP322943100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 23441-75-0
Molekylformel C14H10N2O
Molekylvikt (g/mol) 222.25
Synonym 4-Phenylquinazolin-2(1H)-One
IUPAC-namn 4-phenylquinazolin-2(1H)-one
LEDER O=C1N=C(C2=C(N1)C=CC=C2)C3=CC=CC=C3
Specifikationer
Kemiskt namn eller material 4-phenyl-2,3-dihydroquinazolin-2-one
Kvantitet 100 mg
InChI formel InChI=1S/C14H10N2O/c17-14-15-12-9-5-4-8-11(12)13(16-14)10-6-2-1-3-7-10/h1-9H,(H,15,16,17)
Formel vikt 22.08