missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-5-(trifluoromethyl)thiophen-3-amine, ≥97%, Thermo Scientific™

Catalog Number p-3110698
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
256427-77-7
C11H8F3NS
243.247
MFCD01934528
KBIPYEXWOYDDDL-UHFFFAOYSA-N
4-phenyl-5-trifluoromethyl thiophen-3-amine, 4-phenyl-5-trifluoromethyl thien-3-ylamine, 3-amino-4-phenyl-5-trifluoromethyl thiophene, maybridge3_006438, 4-phenyl-5-trifluoromethyl-3-thienylamine, 4-phenyl-5-trifluoromethyl thiophene-3-amine
2782905
4-phenyl-5-(trifluoromethyl)thiophen-3-amine
C1=CC=C(C=C1)C2=C(SC=C2N)C(F)(F)F
This item is not returnable. View return policy
256427-77-7
C11H8F3NS
243.247
MFCD01934528
KBIPYEXWOYDDDL-UHFFFAOYSA-N
4-phenyl-5-trifluoromethyl thiophen-3-amine, 4-phenyl-5-trifluoromethyl thien-3-ylamine, 3-amino-4-phenyl-5-trifluoromethyl thiophene, maybridge3_006438, 4-phenyl-5-trifluoromethyl-3-thienylamine, 4-phenyl-5-trifluoromethyl thiophene-3-amine
2782905
4-phenyl-5-(trifluoromethyl)thiophen-3-amine
C1=CC=C(C=C1)C2=C(SC=C2N)C(F)(F)F
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 243.25
Percent Purity 97%
Chemical Name or Material 4-Phenyl-5-(trifluoromethyl)thiophen-3-amine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'