Läs mer
4-(Phenylamino)-1-benzyl-4-piperidinecarbonitrile, TRC
Produktkod. 30423141
Change view
Kvantitet:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30423141 500 mg
30418183 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30423141 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP307480500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 968-86-5
Molekylformel C19 H21 N3
Molekylvikt (g/mol) 291.39
Synonym 4-Piperidinecarbonitrile, 4-(phenylamino)-1-(phenylmethyl)-, Isonipecotonitrile, 4-anilino-1-benzyl- (7CI,8CI), 4-(Phenylamino)-1-(phenylmethyl)-4-piperidinecarbonitrile, 1-Benzyl-4-(phenylamino)piperidine-4-carbonitrile, 1-Benzyl-4-cyano-4-anilinopiperidine, 1-Benzyl-4-phenylamino-4-cyanopiperidine, 4-(Phenylamino)-1-(phenylmethyl)-4-piperidinenitrile, 4-Anilino-4-cyano-1-benzylpiperidine, NSC 73005, 1-Benzyl-4-(phenylamino)piperidine-4-carbonitrile, Remifentanil Hydrochloride Imp. H (EP)
IUPAC-namn 4-anilino-1-benzylpiperidine-4-carbonitrile
LEDER N#CC1(CCN(Cc2ccccc2)CC1)Nc3ccccc3
Specifikationer
Kemiskt namn eller material 4-(Phenylamino)-1-benzyl-4-piperidinecarbonitrile
Kvantitet 500 mg
InChI formel InChI=1S/C19H21N3/c20-16-19(21-18-9-5-2-6-10-18)11-13-22(14-12-19)15-17-7-3-1-4-8-17/h1-10,21H,11-15H2
Formel vikt 291.1735