Läs mer
4-Phenylbutylamine, TRC
Produktkod. 30419690
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30419690 500 mg
1 options
Produktkod. 30419690 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP399615500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Phenylbutylamine
CAS 13214-66-9
Molekylformel C10 H15 N
Kvantitet 500 mg
Synonym Primidone Imp. C (EP), Primidone USP Related Compound C, Primidone USP RC C, 1-Amino-4-phenylbutane, 4-Phenyl-1-aminobutane, 4-Phenyl-1-butylamine, 4-Phenyl-n-butylamine, 4-Phenylbutan-1-amine, 4-Phenylbutanamine, Benzenebutanamine, Δ-Phenylbutylamine, Δ-Phenyl-n-butylamine
InChI formel InChI=1S/C10H15N/c11-9-5-4-8-10-6-2-1-3-7-10/h1-3,6-7H,4-5,8-9,11H2
LEDER NCCCCc1ccccc1
IUPAC-namn 4-phenylbutan-1-amine
Molekylvikt (g/mol) 149.23
Formel vikt 149.1204