Läs mer
4-(phenylsulfanyl)phenol, TRC
Produktkod. 30333291
Change view
Kvantitet:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30333291 1 g
30331590 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30333291 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB4816001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-(phenylsulfanyl)phenol
CAS 5633-55-6
Molekylformel C12H10OS
Kvantitet 1 g
InChI formel InChI=1S/C12H10OS/c13-10-6-8-12(9-7-10)14-11-4-2-1-3-5-11/h1-9,13H
LEDER Oc1ccc(Sc2ccccc2)cc1
IUPAC-namn 4-phenylsulfanylphenol
Molekylvikt (g/mol) 202.27
Formel vikt 202.0452