Läs mer
4-Phospho D-Erythronate, TRC
Produktkod. 17902225
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17902225 1 mg
17932735 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17902225 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP3550001MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 57229-25-1
Molekylformel C4H9O8P
Molekylvikt (g/mol) 216.08
Synonym (2R,3R)-2,3-Dihydroxy-4-(phosphonooxy)butanoic Acid, [R-(R*,R*)]-2,3-Dihydroxy-4-(phosphonooxy)butanoic Acid;
LEDER C([C@H]([C@H](C(=O)O)O)O)OP(=O)(O)O
Specifikationer
Kemiskt namn eller material 4-Phospho D-Erythronate
Kvantitet 1 mg
InChI formel InChI=1S/C4H9O8P/c5-2(3(6)4(7)8)1-12-13(9,10)11/h2-3,5-6H,1H2,(H,7,8)(H2,9,10,11)/t2-,3-/m1/s1
Formel vikt 216.0035