missing translation for 'onlineSavingsMsg'
Learn More

4-Phthalimidoacetoacetic Acid Ethyl Ester, TRC

Product Code. 30417514
Change view
Click to view available options
Kvantitet:
2.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30417514 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30417514 Supplier Toronto Research Chemicals Supplier No. TRCP3846352.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 4-Phthalimidoacetoacetic Acid Ethyl Ester
CAS 13855-80-6
Molekylformel C14H13NO5
Kvantitet 2.5 mg
Synonym Ethyl (phthalimidoacetyl)acetate, Ethyl 4-(1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)-3-oxobutanoate, Ethyl 4-phthalimidoacetoacetate, 4-Phthalimidoacetoacetic acid ethyl ester, 2-Isoindolineacetoacetic acid','1,3-dioxo-','ethyl ester (7CI','8CI), Ethyl 1,3-dihydro-β,1,3-trioxo-2H-isoindole-2-butanoate (ACI), Ethyl (phthalimidoacetyl)acetate, Ethyl 4-(1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)-3-oxobutanoate, Ethyl 4-phthalimidoacetoacetate
InChI formel InChI=1S/C14H13NO5/c1-2-20-12(17)7-9(16)8-15-13(18)10-5-3-4-6-11(10)14(15)19/h3-6H,2,7-8H2,1H3
LEDER O=C(OCC)CC(=O)CN1C(=O)C=2C=CC=CC2C1=O
IUPAC-namn 2H-Isoindole-2-butanoic acid','1,3-dihydro-β,1,3-trioxo-','ethyl ester
Molekylvikt (g/mol) 275.26
Formel vikt 275.08
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.