Läs mer
4-Phthalimidoacetoacetic Acid Ethyl Ester, TRC
Produktkod. 30428168
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30428168 5 mg
1 options
Produktkod. 30428168 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP3846355MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Phthalimidoacetoacetic Acid Ethyl Ester
CAS 13855-80-6
Molekylformel C14H13NO5
Kvantitet 5 mg
Synonym Ethyl (phthalimidoacetyl)acetate, Ethyl 4-(1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)-3-oxobutanoate, Ethyl 4-phthalimidoacetoacetate, 4-Phthalimidoacetoacetic acid ethyl ester, 2-Isoindolineacetoacetic acid','1,3-dioxo-','ethyl ester (7CI','8CI), Ethyl 1,3-dihydro-β,1,3-trioxo-2H-isoindole-2-butanoate (ACI), Ethyl (phthalimidoacetyl)acetate, Ethyl 4-(1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)-3-oxobutanoate, Ethyl 4-phthalimidoacetoacetate
InChI formel InChI=1S/C14H13NO5/c1-2-20-12(17)7-9(16)8-15-13(18)10-5-3-4-6-11(10)14(15)19/h3-6H,2,7-8H2,1H3
LEDER O=C(OCC)CC(=O)CN1C(=O)C=2C=CC=CC2C1=O
IUPAC-namn 2H-Isoindole-2-butanoic acid','1,3-dihydro-β,1,3-trioxo-','ethyl ester
Molekylvikt (g/mol) 275.26
Formel vikt 275.08