missing translation for 'onlineSavingsMsg'
Learn More

4-(piperidinomethyl)benzoic acid, 97%, Thermo Scientific™

Catalog Number p-4510673
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
159691-33-5
C13H17NO2
219.284
RNCCMTVMMFUIKR-UHFFFAOYSA-N
4-piperidin-1-ylmethyl benzoic acid, 4-piperidin-1-ylmethyl-benzoic acid, benzoic acid,4-1-piperidinylmethyl, 4-piperidylmethyl benzoic acid, 4-piperidinomethyl benzoic acid, 4-piperidin-1-yl methyl benzoic acid
1092975
4-(piperidin-1-ylmethyl)benzoic acid
C1CCN(CC1)CC2=CC=C(C=C2)C(=O)O
This item is not returnable. View return policy
159691-33-5
C13H17NO2
219.284
RNCCMTVMMFUIKR-UHFFFAOYSA-N
4-piperidin-1-ylmethyl benzoic acid, 4-piperidin-1-ylmethyl-benzoic acid, benzoic acid,4-1-piperidinylmethyl, 4-piperidylmethyl benzoic acid, 4-piperidinomethyl benzoic acid, 4-piperidin-1-yl methyl benzoic acid
1092975
4-(piperidin-1-ylmethyl)benzoic acid
C1CCN(CC1)CC2=CC=C(C=C2)C(=O)O
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 219.28
Percent Purity 97%
Chemical Name or Material 4-(Piperidinomethyl)benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'