Learn More
Gilac™ 4 Pivoting Wheel Dolly
Product Code. 30245714
Change view
Click to view available options
Material:
ABS Plastic, Chrome-plated Steel
Unit Size:
Each
Product Code. Material Unit Size
30245714 ABS Plastic, Chrome-plated Steel Each
1 options
Product Code. 30245714 Supplier Gilac™ Supplier No. G170109
  • ONLINE EXCLUSIVE PRICE
    1310.84 SEK
    731.66 SEK / Paket med 1
    Save 579.18 SEK
    44% Av
Beräknad leverans: 09-12-2026
Universal dolly on wheels. Excellent stability and great manoeuvrability thanks to its 4 ball bearing mounted drive wheels.
  • Excellent stability and great manoeuvrability thanks to its 4 ball bearing mounted drive wheels
  • 250 kg load capacity
  • Outstanding quality, outclassing usual industry standards
  • ABS injected base with solid frame guaranteeing unrivalled durability
  • 100 mm black PP wheels
  • GILAC removable pulling bar available (as an option)
  • Compatible with all GILAC 600 x 400 mm and 400 x 300 mm containers and crates
  • Dolly also available in black ABS-PC
Specifications
Type Dolly
Holds Dolly
Material ABS Plastic, Chrome-plated Steel