Läs mer
4-(Propylthio)-1,2-benzenediamine, TRC
Produktkod. 30423015
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30423015 1 g
30412878 500 mg
30410907 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30423015 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP8384701G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 66608-52-4
Molekylformel C9H14N2S
Molekylvikt (g/mol) 182.29
Synonym 4-(Propylthio)-1,2-phenylenediamine, 4-Propylthio-o-phenylenediamine
IUPAC-namn 4-propylsulfanylbenzene-1,2-diamine
LEDER CCCSc1ccc(N)c(N)c1
Specifikationer
Kemiskt namn eller material 4-(Propylthio)-1,2-benzenediamine
Kvantitet 1 g
InChI formel InChI=1S/C9H14N2S/c1-2-5-12-7-3-4-8(10)9(11)6-7/h3-4,6H,2,5,10-11H2,1H3
Formel vikt 182.0878