missing translation for 'onlineSavingsMsg'
Learn More

4-Pyrid-4-ylbenzoic acid, 95%, Thermo Scientific™

Catalog Number p-3110720
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
4385-76-6
C12H9NO2
199.21
MFCD04114574
DZLGZIGLHCRIMF-UHFFFAOYSA-N
4-pyridin-4-yl-benzoic acid, 4-4-pyridyl benzoic acid, 4-pyridin-4-yl benzoic acid, 4-pyrid-4-ylbenzoic acid, 4-4-pyridinyl benzoic acid, 4-4-carboxyphenyl pyridine, 4'-pyridyl-4-benzoic acid, benzoic acid, 4-4-pyridinyl, pubchem22345, acmc-1adxv
1520811
OC(=O)C1=CC=C(C=C1)C1=CC=NC=C1
This item is not returnable. View return policy
4385-76-6
C12H9NO2
199.21
MFCD04114574
DZLGZIGLHCRIMF-UHFFFAOYSA-N
4-pyridin-4-yl-benzoic acid, 4-4-pyridyl benzoic acid, 4-pyridin-4-yl benzoic acid, 4-pyrid-4-ylbenzoic acid, 4-4-pyridinyl benzoic acid, 4-4-carboxyphenyl pyridine, 4'-pyridyl-4-benzoic acid, benzoic acid, 4-4-pyridinyl, pubchem22345, acmc-1adxv
1520811
OC(=O)C1=CC=C(C=C1)C1=CC=NC=C1
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 4-pyridin-4-ylbenzoic acid
Formula Weight 199.21
Percent Purity 95%
Chemical Name or Material 4-Pyrid-4-ylbenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'