Learn More
4-Pyridinealdoxime, TRC
Product Code. 30413378
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity Unit Size
30413378 5 g 5g
1 options
Product Code. 30413378 Supplier Toronto Research Chemicals Supplier No. TRCP9913005G
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Pyridinealdoxime
CAS 696-54-8
Molecular Formula C6 H6 N2 O
Quantity 5 g
Synonym 4-Pyridinecarboxaldehyde','oxime, 4-Pyridinecarboxaldehyde oxime, Isonicotinaldehyde','oxime (6CI,8CI), 4-(Hydroxyiminomethyl)pyridine, 4-Pyridinaldoxime, 4-Pyridinealdoxime, NSC 63847, Pyridine-4-carboxaldoxime
InChI Formula InChI=1S/C6H6N2O/c9-8-5-6-1-3-7-4-2-6/h1-5,9H/b8-5+
SMILES O\N=C\c1ccncc1
IUPAC Name (NE)-N-(pyridin-4-ylmethylidene)hydroxylamine
Molecular Weight (g/mol) 122.12
Formula Weight 122.048