missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 4-Pyridineethanesulfonic acid, 99%
Click to view available options
Quantity:
25 g
100 g
Specifications
Melting Point | ∽295°C (decomposition) |
Quantity | 25 g |
UN Number | UN2585 |
Beilstein | 136690 |
Formula Weight | 187.22 |
Percent Purity | 99% |
Chemical Name or Material | 4-Pyridineethanesulfonic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'