Läs mer
4-Pyridoxolactone, TRC
Produktkod. 30413333
Change view
Kvantitet:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30413333 100 mg
30408960 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30413333 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP997213100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4753-19-9
Molekylformel C8H7NO3
Molekylvikt (g/mol) 165.15
Synonym 4-Pyridoxic Acid Lactone, beta-Pyracin, beta-Pyracine, 3-Hydroxy-5-(hydroxymethyl)-2-methyl-4-pyridinecarboxylic Acid gamma-Lactone, 3-Hydroxy-5-(hydroxymethyl)-2-methyl-isonicotinic Acid gamma-Lactone, 7-Hydroxy-6-methyl-furo[3,4-c]pyridin-1(3H)-one
IUPAC-namn 7-hydroxy-6-methyl-3H-furo[3,4-c]pyridin-1-one
LEDER Cc1ncc2COC(=O)c2c1O
Specifikationer
Kemiskt namn eller material 4-Pyridoxolactone
Kvantitet 100 mg
InChI formel InChI=1S/C8H7NO3/c1-4-7(10)6-5(2-9-4)3-12-8(6)11/h2,10H,3H2,1H3
Formel vikt 165.0426