missing translation for 'onlineSavingsMsg'
Läs mer

4-Pyridylhydrazine Hydrochloride, TRC

Produktkod. 30421920
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30421920 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30421920 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP997293500MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 4-Pyridylhydrazine Hydrochloride
CAS 52834-40-9
Molekylformel C5 H7 N3 . H Cl
Kvantitet 500 mg
Synonym Pyridine','4-hydrazino-','hydrochloride (7CI,9CI), 4-Hydrazinopyridine hydrochloride
InChI formel InChI=1S/C5H7N3.ClH/c6-8-5-1-3-7-4-2-5;/h1-4H,6H2,(H,7,8);1H
LEDER Cl.NNc1ccncc1
IUPAC-namn pyridin-4-ylhydrazine;hydrochloride
Molekylvikt (g/mol) 145.59
Formel vikt 145.041
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.