missing translation for 'onlineSavingsMsg'
Learn More

4-Pyrimidin-2-ylbenzonitrile, 95%, Thermo Scientific™

Catalog Number p-3541173
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
78322-96-0
C11H7N3
181.198
MFCD09817491
WGCKVXHOTDDNOA-UHFFFAOYSA-N
4-pyrimidin-2-yl benzonitrile, 4-cyanophenyl pyrimidine, 2-4-cyanophenyl pyrimidine, 4-pyrimidin-2-yl-benzonitrile, 4-2-pyrimidinyl benzonitrile, benzonitrile, 4-2-pyrimidinyl, 4-pyrimidin-2-ylbenzenecarbonitrile
15486310
4-pyrimidin-2-ylbenzonitrile
C1=CN=C(N=C1)C2=CC=C(C=C2)C#N
This item is not returnable. View return policy
78322-96-0
C11H7N3
181.198
MFCD09817491
WGCKVXHOTDDNOA-UHFFFAOYSA-N
4-pyrimidin-2-yl benzonitrile, 4-cyanophenyl pyrimidine, 2-4-cyanophenyl pyrimidine, 4-pyrimidin-2-yl-benzonitrile, 4-2-pyrimidinyl benzonitrile, benzonitrile, 4-2-pyrimidinyl, 4-pyrimidin-2-ylbenzenecarbonitrile
15486310
4-pyrimidin-2-ylbenzonitrile
C1=CN=C(N=C1)C2=CC=C(C=C2)C#N
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 181.2
Percent Purity 95%
Chemical Name or Material 4-Pyrimidin-2-ylbenzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'