Learn More
4-Pyrimidin-2-ylbenzoyl chloride, TECH, Thermo Scientific™
Product Code. 10679233
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10679233 250 mg Amber Glass Bottle 250mg
10613065 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10679233 Supplier Thermo Scientific Maybridge Supplier No. CC56202CB
 Restricted Item
Beräknad leverans: 09-12-2026
Add to basket
Chemical Identifiers
CAS 679806-84-9
Molecular Formula C11H7ClN2O
Molecular Weight (g/mol) 218.64
MDL Number MFCD09064974
InChI Key AQFRTZZTRGGBKO-UHFFFAOYSA-N
Synonym 4-pyrimidin-2-yl benzoyl chloride, benzoyl chloride,4-2-pyrimidinyl, 4-pyrimidin-2-yl-benzoyl chloride
PubChem CID 21092945
IUPAC Name 4-pyrimidin-2-ylbenzoyl chloride
SMILES C1=CN=C(N=C1)C2=CC=C(C=C2)C(=O)Cl
Specifications
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 218.64
Percent Purity 90%
Chemical Name or Material 4-Pyrimidin-2-ylbenzoyl chloride