Learn More
4-(Pyrrolidin-1-ylmethyl)aniline, 97%, Thermo Scientific™
Product Code. 10633245
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10633245 250 mg 250mg
10183224 1 g 1g
10336384 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10633245 Supplier Thermo Scientific Maybridge Supplier No. CC55614CB
 Restricted Item
Beräknad leverans: 11-12-2026
Add to basket
Chemical Identifiers
CAS 142335-64-6
Molecular Formula C11H17N2
Molecular Weight (g/mol) 177.27
MDL Number MFCD03724739
InChI Key SFEAIUCOZWDYMJ-UHFFFAOYSA-O
Synonym 4-pyrrolidin-1-ylmethyl aniline, 4-pyrrolidin-1-ylmethyl-phenylamine, benzenamine, 4-1-pyrrolidinylmethyl, 4-1-pyrrolidinylmethyl aniline, 4-pyrrolidin-1-ylmethyl-aniline, 4-pyrrolidinylmethyl phenylamine, 1-4-aminobenzyl pyrrolidine, 4-pyrrolizinomethyl aniline, 4-pyrrolidinyl-methyl aniline, cbi-bb zero/006543
PubChem CID 776852
IUPAC Name 4-(pyrrolidin-1-ylmethyl)aniline
SMILES NC1=CC=C(C[NH+]2CCCC2)C=C1
Specifications
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 176.26
Chemical Name or Material 4-(Pyrrolidin-1-ylmethyl)aniline