missing translation for 'onlineSavingsMsg'
Learn More

4-(t-Boc-amino)-1-butyl Bromide, TRC

Product Code. 30346281
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Kvantitet:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30346281 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30346281 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCB6015255G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 4-(t-Boc-amino)-1-butyl Bromide
CAS 164365-88-2
Molekylformel C9 H18 Br N O2
Kvantitet 5 g
InChI formel InChI=1S/C9H18BrNO2/c1-9(2,3)13-8(12)11-7-5-4-6-10/h4-7H2,1-3H3,(H,11,12)
LEDER CC(C)(C)OC(=O)NCCCCBr
IUPAC-namn tert-butyl N-(4-bromobutyl)carbamate
Molekylvikt (g/mol) 252.15
Formel vikt 251.0521
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.