Läs mer
4-(tert-Butoxycarbonyloxy)benzylalcohol, TRC
Produktkod. 30335261
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30335261 100 mg
1 options
Produktkod. 30335261 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB690355100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-(tert-Butoxycarbonyloxy)benzylalcohol
CAS 156281-11-7
Molekylformel C12H16O4
Kvantitet 100 mg
Synonym tert-Butyl p-(Hydroxymethyl)phenyl Carbonate, Carbonic acid 1,1-Dimethylethyl 4-(Hydroxymethyl)phenyl Ester
InChI formel InChI=1S/C12H16O4/c1-12(2,3)16-11(14)15-10-6-4-9(8-13)5-7-10/h4-7,13H,8H2,1-3H3
LEDER CC(C)(C)OC(=O)Oc1ccc(CO)cc1
IUPAC-namn tert-butyl [4-(hydroxymethyl)phenyl] carbonate
Molekylvikt (g/mol) 224.25
Formel vikt 224.1049