Läs mer
4-tert-Butyl-1,2-bis(chloromethyl)benzene, TRC
Produktkod. 30348442
Change view
Kvantitet:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30348442 100 mg
30339780 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30348442 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB807070100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-tert-Butyl-1,2-bis(chloromethyl)benzene
CAS 141788-33-2
Molekylformel C12H16Cl2
Kvantitet 100 mg
Synonym 1,2-bis(Chloromethyl)-4-(1,1-dimethylethyl)-benzene
InChI formel InChI=1S/C12H16Cl2/c1-12(2,3)11-5-4-9(7-13)10(6-11)8-14/h4-6H,7-8H2,1-3H3
LEDER CC(C)(C)c1ccc(CCl)c(CCl)c1
IUPAC-namn 4-tert-butyl-1,2-bis(chloromethyl)benzene
Molekylvikt (g/mol) 231.16
Formel vikt 230.0629