Läs mer
4-tert-Butyl-2-chlorophenol (Technical Grade), TRC
Produktkod. 30334039
Change view
Click to view available options
Kvantitet:
100 mg
Product Code. Kvantitet
30334039 100 mg
1 options
Product Code. 30334039 Supplier Toronto Research Chemicals Supplier No. TRCB807038100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material 4-tert-Butyl-2-chlorophenol
CAS 98-28-2
Molekylformel C10 H13 Cl O
Kvantitet 100 mg
Synonym Phenol, 4-tert-butyl-2-chloro- (6CI,7CI,8CI), 2-Chloro-4-(1,1-dimethylethyl)phenol, 2-Chloro-4-tert-butylphenol, 4-tert-Butyl-2-chlorophenol, NSC 8464
InChI formel InChI=1S/C10H13ClO/c1-10(2,3)7-4-5-9(12)8(11)6-7/h4-6,12H,1-3H3
LEDER CC(C)(C)c1ccc(O)c(Cl)c1
IUPAC-namn 4-tert-butyl-2-chlorophenol
Molekylvikt (g/mol) 184.66
Formel vikt 184.0655