missing translation for 'onlineSavingsMsg'
Learn More

4-(tert-Butyl)benzylchloride, 97%, Thermo Scientific Chemicals

Catalog Number 10737962
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
19692-45-6
C11H15Cl
182.69
MFCD00000918
WAXIFMGAKWIFDQ-UHFFFAOYSA-N
4-tert-butylbenzyl chloride, 1-tert-butyl-4-chloromethyl benzene, p-tert-butylbenzyl chloride, 4-tert-butyl benzyl chloride, benzene, 1-chloromethyl-4-1,1-dimethylethyl, 4-tert-butyl benzylchloride, p-t-butylbenzyl chloride, 1-chloromethyl-4-tert-butylbenzene, p-tert-butyl-.alpha.-chlorotoluene
88198
1-tert-butyl-4-(chloromethyl)benzene
CC(C)(C)C1=CC=C(CCl)C=C1
This item is not returnable. View return policy
19692-45-6
C11H15Cl
182.69
MFCD00000918
WAXIFMGAKWIFDQ-UHFFFAOYSA-N
4-tert-butylbenzyl chloride, 1-tert-butyl-4-chloromethyl benzene, p-tert-butylbenzyl chloride, 4-tert-butyl benzyl chloride, benzene, 1-chloromethyl-4-1,1-dimethylethyl, 4-tert-butyl benzylchloride, p-t-butylbenzyl chloride, 1-chloromethyl-4-tert-butylbenzene, p-tert-butyl-.alpha.-chlorotoluene
88198
1-tert-butyl-4-(chloromethyl)benzene
CC(C)(C)C1=CC=C(CCl)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9400g/mL
Boiling Point 98°C to 103°C (7.0 mmHg)
Flash Point 94°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CC6H4CH2Cl
Refractive Index 1.5205 to 1.5225
Quantity 5 g
Specific Gravity 0.94
Formula Weight 182.69
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-(tert-Butyl)benzylchloride, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 4-(tert-Butyl)benzylchloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'