Läs mer
4-tert-Octylphenol-13C6, TRC
Produktkod. 17931715
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17931715 10 mg
17973605 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17931715 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO29378210MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1173020-24-0
Molekylformel 13C6 C8 H22 O
Molekylvikt (g/mol) 212.28
Synonym 4-tert-Octylphenol 13C6 (phenyl 13C6)
IUPAC-namn 4-(2,4,4-trimethylpentan-2-yl)(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-trien-1-ol
LEDER CC(C)(C)CC(C)(C)[13c]1[13cH][13cH][13c](O)[13cH][13cH]1
Specifikationer
Kemiskt namn eller material 4-tert-Octylphenol ring-13C6
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C14H22O/c1-13(2,3)10-14(4,5)11-6-8-12(15)9-7-11/h6-9,15H,10H2,1-5H3/i6+1,7+1,8+1,9+1,11+1,12+1
Formel vikt 212.1872
Procent renhet >95%