Läs mer
4-tert-Octylphenol Diethoxylate-13C6, TRC
Produktkod. 30009378
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009378 10 mg
30006797 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30009378 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO29379710MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1173020-69-3
Molekylformel C1213C6H30O3
Molekylvikt (g/mol) 300.38
Synonym 2-[2-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]ethoxy]ethanol-13C6, 2-[2-[p-(1,1,3,3-Tetramethylbutyl)phenoxy]ethoxy]ethanol-13C6, OP2EO-13C6
IUPAC-namn 2-[2-[4-(2,4,4-trimethylpentan-2-yl)(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-trien-1-yl]oxyethoxy]ethanol
LEDER OCCOCCO[13C]1=[13CH][13CH]=[13C](C(CC(C)(C)C)(C)C)[13CH]=[13CH]1
Specifikationer
Kemiskt namn eller material 4-tert-Octylphenol Diethoxylate-13C6
Kvantitet 10 mg
InChI formel InChI=1S/C18H30O3/c1-17(2,3)14-18(4,5)15-6-8-16(9-7-15)21-13-12-20-11-10-19/h6-9,19H,10-14H2,1-5H3/i6+1,7+1,8+1,9+1,15+1,16+1
Formel vikt 300.2396