Läs mer
4-tert-Octylphenol Monoethoxylate-13C6, TRC
Produktkod. 30009229
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009229 10 mg
30007868 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30009229 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO29384210MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1173019-48-1
Molekylformel 13C6 C10 H26 O2
Molekylvikt (g/mol) 256.33
Synonym 4-tert-Octylphenol-mono-ethoxylate 13C6, 4-tert-Octylphenol monoethoxylate (ring 13C6), Ethanol, 2-[4-(1,1,3,3-tetramethylbutyl)phenoxy-1,2,3,4,5,6-13C6]-, Ethanol, 2-[4-(1,1,3,3-tetramethylbutyl)phenoxy-1,2,3,4,5,6-13C6]-
IUPAC-namn 2-[4-(2,4,4-trimethylpentan-2-yl)(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-trien-1-yl]oxyethanol
LEDER CC(C)(C)CC(C)(C)[13c]1[13cH][13cH][13c](OCCO)[13cH][13cH]1
Specifikationer
Kemiskt namn eller material 4-tert-Octylphenol Monoethoxylate-13C6
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C16H26O2/c1-15(2,3)12-16(4,5)13-6-8-14(9-7-13)18-11-10-17/h6-9,17H,10-12H2,1-5H3/i6+1,7+1,8+1,9+1,13+1,14+1
Formel vikt 256.2134
Procent renhet >95%