Läs mer
4-tert-Octylphenol, TRC
Produktkod. 30012708
Change view
Kvantitet:
1 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012708 25 g
30009788 10 g
17913305 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30012708 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO29378025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 140-66-9
Molekylformel C14 H22 O
Molekylvikt (g/mol) 206.32
Synonym 4-tert-Octylphenol, p-(1,1,3,3-Tetramethylbutyl)phenol, p-Octylphenol, p-t-Octylphenol, p-tert-Octylphenol, Phenol, 4-(1,1,3,3-tetramethylbutyl)- (9CI, ACI), 4-(1,1,3,3-Tetramethylbutyl)phenol (ACI), Phenol, p-(1,1,3,3-tetramethylbutyl)- (6CI, 8CI), 4-(2,2,4-Trimethylpentan-2-yl)phenol, 4-(2,4,4-Trimethylpentan-2-yl)phenol, 4-Octylphenol, 4-tert-Octylphenol, NSC 5427, NSC 7248
IUPAC-namn 4-(2,4,4-trimethylpentan-2-yl)phenol
LEDER CC(C)(C)CC(C)(C)c1ccc(O)cc1
Specifikationer
Kemiskt namn eller material 4-tert-Octylphenol
Kvantitet 25 g
InChI formel InChI=1S/C14H22O/c1-13(2,3)10-14(4,5)11-6-8-12(15)9-7-11/h6-9,15H,10H2,1-5H3
Formel vikt 206.1671