Learn More
4-(Thien-3-yl)benzoic acid, 97%, Thermo Scientific™
Catalog Number p-7077343
Click to view available options
Quantity:
1 g
5 g
29886-64-4
C11H8O2S
204.25
FISAUHGRILVMDP-UHFFFAOYSA-N
4-3-thienyl benzoic acid, 4-thiophen-3-yl benzoic acid, 4-thiophen-3-yl-benzoic acid, benzoic acid, 4-3-thienyl, 4-thien-3-ylbenzoic acid, 4-3-thienyl-benzoic acid, maybridge3_004545, acmc-1cisg
736862
4-thiophen-3-ylbenzoic acid
C1=CC(=CC=C1C2=CSC=C2)C(=O)O
29886-64-4
C11H8O2S
204.25
FISAUHGRILVMDP-UHFFFAOYSA-N
4-3-thienyl benzoic acid, 4-thiophen-3-yl benzoic acid, 4-thiophen-3-yl-benzoic acid, benzoic acid, 4-3-thienyl, 4-thien-3-ylbenzoic acid, 4-3-thienyl-benzoic acid, maybridge3_004545, acmc-1cisg
736862
4-thiophen-3-ylbenzoic acid
C1=CC(=CC=C1C2=CSC=C2)C(=O)O
Melting Point 281.0°C to 283.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 204.25
Percent Purity 97%
Chemical Name or Material 4-(Thien-3-yl)benzoic acid
compliance-icons
Product Identifier
  • 4-(Thien-3-yl)benzoic acid, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.