Läs mer
4-(Trifluoromethoxy)benzoic Acid, TRC
Produktkod. 30454727
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30454727 5 g
1 options
Produktkod. 30454727 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT7907205G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-(Trifluoromethoxy)benzoic Acid
CAS 330-12-1
Molekylformel C8H5F3O3
Kvantitet 5 g
Synonym alpha,alpha,alpha-Trifluoro-p-anisic Acid, p-(Trifluoromethoxy)benzoic Acid
InChI formel InChI=1S/C8H5F3O3/c9-8(10,11)14-6-3-1-5(2-4-6)7(12)13/h1-4H,(H,12,13)
LEDER OC(=O)c1ccc(OC(F)(F)F)cc1
IUPAC-namn 4-(trifluoromethoxy)benzoic acid
Molekylvikt (g/mol) 206.12
Formel vikt 206.0191