missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Trifluoromethyl-o-phenylenediamine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
3,4-Diaminobenzotrifluoride is commonly used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,4-Diaminobenzotrifluoride is commonly used as pharmaceutical intermediate.
Solubility
Miscible with water.
Notes
Stable at room temperature in closed containers under normal storage and handling conditions. Incompatible to oxidizing agents. Store in a cool, dry place.
3,4-Diaminobenzotrifluoride is commonly used as pharmaceutical intermediate.
Solubility
Miscible with water.
Notes
Stable at room temperature in closed containers under normal storage and handling conditions. Incompatible to oxidizing agents. Store in a cool, dry place.
Specifications
Melting Point | 58°C to 62°C |
Quantity | 1 g |
UN Number | UN2811 |
Beilstein | 909258 |
Solubility Information | Miscible with water. |
Formula Weight | 176.14 |
Percent Purity | 98% |
Chemical Name or Material | 4-Trifluoromethyl-o-phenylenediamine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'