Läs mer
4-Vinylbenzyl Bromide (stabilized with TBC, ~90%), TRC
Produktkod. 30456811
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30456811 100 mg
30449781 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30456811 Leverantör Toronto Research Chemicals Leverantörsnummer TRCV371010100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13368-25-7
Molekylformel C9H9Br
Molekylvikt (g/mol) 197.07
Synonym 1-(Bromomethyl)-4-ethenyl-benzene, p-(Bromomethyl)-styrene, 1-Bromomethyl-4-vinylbenzene, 4-(Bromomethyl)styrene, p-(Bromomethyl)styrene, p-Vinylbenzyl bromide
IUPAC-namn 1-(bromomethyl)-4-vinylbenzene
LEDER BrCC1=CC=C(C=C)C=C1
Specifikationer
Kemiskt namn eller material 4-Vinylbenzyl Bromide
Kvantitet 100 mg
InChI formel InChI=1S/C9H9Br/c1-2-8-3-5-9(7-10)6-4-8/h2-6H,1,7H2
Formel vikt 195.99