Läs mer
4-Vinylsyringol, TRC
Produktkod. 30240790
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30240790 250 mg
30240554 50 mg
30240858 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30240790 Leverantör Toronto Research Chemicals Leverantörsnummer TRCV280000250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 28343-22-8
Molekylformel C10 H12 O3
Molekylvikt (g/mol) 180.2
Synonym 4-Ethenyl-2,6-Dimethoxyphenol, 2,6-Dimethoxy-4-Ethenylphenol, 2,6-Dimethoxy-4-Vinylphenol, 3,5-Dimethoxy-4-Hydroxystyrene, 4-Vinyl-2,6-Dimethoxyphenol, Canolol, Syringlyethene, Vinylsyringol
IUPAC-namn 4-ethenyl-2,6-dimethoxyphenol
LEDER Coc1Cc(C=C)Cc(Oc)C1O
Specifikationer
Kemiskt namn eller material 4-Vinylsyringol
Kvantitet 250 mg
InChI formel InChI=1S/C10H12O3/c1-4-7-5-8(12-2)10(11)9(6-7)13-3/h4-6,11H,1H2,2-3H3
Formel vikt 180.0786