Learn More
4-Vinylsyringol, TRC
Product Code. 30240858
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30240858 500 mg 500mg
30240790 250 mg 250mg
30240554 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30240858 Supplier Toronto Research Chemicals Supplier No. TRCV280000500MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 28343-22-8
Molecular Formula C10 H12 O3
Molecular Weight (g/mol) 180.2
Synonym 4-Ethenyl-2,6-Dimethoxyphenol, 2,6-Dimethoxy-4-Ethenylphenol, 2,6-Dimethoxy-4-Vinylphenol, 3,5-Dimethoxy-4-Hydroxystyrene, 4-Vinyl-2,6-Dimethoxyphenol, Canolol, Syringlyethene, Vinylsyringol
IUPAC Name 4-ethenyl-2,6-dimethoxyphenol
SMILES Coc1Cc(C=C)Cc(Oc)C1O
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C10H12O3/c1-4-7-5-8(12-2)10(11)9(6-7)13-3/h4-6,11H,1H2,2-3H3
Formula Weight 180.0786
Chemical Name or Material 4-Vinylsyringol